siRNA Details
Gene Symbol: FAM9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174901.5 | NP_777561.1 | 8 | 1108 | ||
XM_005274460.2 | XP_005274517.1 | 7 | 2912 | |||
XM_017029321.1 | XP_016884810.1 | 6 | 3246 | |||
GenBank mRNA | AF494345.1 | - | 8 | 991 | ||
BX648111.1 | - | 6 | 5711 | |||
BX648919.1 | - | 6 | 5712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
