siRNA Details
Gene Symbol: TMEM26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134508.1 | 6 | 1098 | |||
GenBank mRNA | BC042872.1 | - | 6 | 815 | 5164050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
