siRNA Details
Gene Symbol: STXBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721797.3 | XP_006721860.1 | 12 | 1310 | ||
XM_006721798.3 | XP_006721861.1 | 12 | 1301 | |||
XM_017024414.1 | XP_016879903.1 | 13 | 1360 | |||
GenBank mRNA | BC041485.2 | - | 11 | 1113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
