siRNA Details
Gene Symbol: JAKMIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105521.3 | NP_001098991.1 | 11 | 1824 | ||
NM_001323086.1 | NP_001310015.1 | 11 | 1824 | |||
NM_001323087.1 | NP_001310016.1 | 11 | 1824 | |||
NM_001323088.1 | NP_001310017.1 | 11 | 1818 | |||
NM_001323090.1 | NP_001310019.1 | 11 | 1824 | |||
XM_005252674.4 | XP_005252731.1 | 11 | 3818 | |||
XM_005252676.4 | XP_005252733.1 | 10 | 3596 | |||
XM_011539676.2 | XP_011537978.1 | 11 | 2070 | |||
XM_011539682.2 | XP_011537984.1 | 10 | 3602 | |||
XM_017016095.1 | XP_016871584.1 | 11 | 3818 | |||
XM_017016096.1 | XP_016871585.1 | 11 | 2069 | |||
XM_017016097.1 | XP_016871586.1 | 10 | 3596 | |||
XM_017016098.1 | XP_016871587.1 | 11 | 3818 | |||
GenBank mRNA | AK126964.1 | - | 1 | 111 |
