siRNA Details
Gene Symbol: ATP5L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165877.1 | NP_001159349.1 | 1 | 651 | ||
GenBank mRNA | AF092923.1 | - | 1 | 651 | ||
BC093719.1 | - | 1 | 361 | |||
BC093721.1 | - | 1 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
