siRNA Details
Gene Symbol: NR2C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI986459.1 | - | 1 | 50 |
Gene Symbol: MRPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022497.4 | NP_071942.1 | 4 | 3872 | ||
GenBank mRNA | AK024433.1 | - | 4 | 3787 | ||
AK294123.1 | - | 4 | 1097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
