siRNA Details
Gene Symbol: EBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022659.3 | NP_073150.2 | 16 | 2157 | ||
GenBank mRNA | AK021562.1 | - | 9 | 1196 | ||
AK304372.1 | - | 15 | 1805 | |||
BC069665.1 | - | 9 | 1174 | |||
BC069726.1 | - | 9 | 1177 | |||
BC069747.1 | - | 9 | 1176 | |||
BC069768.1 | - | 9 | 1177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
