siRNA Details
Gene Symbol: ZNF267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265588.1 | NP_001252517.1 | 4 | 544 | ||
NM_003414.5 | NP_003405.3 | 3 | 393 | |||
NR_049749.1 | 3 | 393 | ||||
XM_017022865.1 | XP_016878354.1 | 4 | 480 | |||
XM_017022866.1 | XP_016878355.1 | 4 | 420 | |||
GenBank mRNA | AF220492.1 | - | 3 | 346 | ||
AK292573.1 | - | 3 | 218 | |||
BC036367.1 | - | 3 | 317 | 4828269 | ||
BC127088.1 | - | 3 | 316 | 40123276 | ||
BC127089.1 | - | 3 | 316 | 40123277 | ||
DA657367.1 | - | 3 | 344 | |||
DB091947.1 | - | 4 | 470 | |||
DB484386.1 | - | 3 | 392 | |||
X78925.1 | - | 2 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
