siRNA Details
Gene Symbol: SOWAHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023016.3 | NP_075392.2 | 1 | 1438 | ||
GenBank mRNA | AK025523.1 | - | 1 | 1407 | ||
BC036652.1 | - | 1 | 1462 | 5267818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
