siRNA Details
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 10 | 940 | ||
XM_006721162.1 | XP_006721225.1 | 9 | 870 | |||
XM_011522928.1 | XP_011521230.1 | 10 | 1118 | |||
XM_011522929.1 | XP_011521231.1 | 10 | 983 | |||
XM_011522930.2 | XP_011521232.1 | 10 | 951 | |||
XM_011522931.2 | XP_011521233.1 | 10 | 951 | |||
XM_017023012.1 | XP_016878501.1 | 10 | 967 | |||
GenBank mRNA | AJ441184.1 | - | 10 | 939 | ||
AK056456.1 | - | 10 | 939 | |||
AK096727.1 | - | 2 | 146 | |||
AK128387.1 | - | 10 | 1036 | |||
AK315590.1 | - | 10 | 943 | |||
BC013735.1 | - | 8 | 726 | |||
BC032542.1 | - | 10 | 946 | 5534376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
