siRNA Details
Gene Symbol: SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204856.1 | NP_001191785.1 | 6, 7 | 781 | ||
NM_024041.3 | NP_076946.1 | 6, 7 | 732 | |||
NR_037937.1 | 6, 7 | 682 | ||||
GenBank mRNA | AK056322.1 | - | 6, 7 | 701 | ||
AV718162.2 | - | 6, 7 | 658 | |||
BC000264.1 | - | 6, 7 | 676 | 3356277 | ||
BU540926.1 | - | 6, 7 | 548 |
Gene Symbol: TNFAIP8L2-SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204848.1 | NP_001191777.1 | 6, 7 | 644 | ||
GenBank mRNA | BG531092.1 | - | 6, 7 | 611 | ||
CR999387.1 | - | 5, 6 | 625 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
