siRNA Details
Gene Symbol: PANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024594.3 | NP_078870.1 | 2 | 499 | ||
GenBank mRNA | AK022961.1 | - | 2 | 244 | ||
AK223285.1 | - | 2 | 228 | |||
BC013705.1 | - | 2 | 473 | 4343270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
