siRNA Details
Gene Symbol: ASB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024701.3 | NP_078977.2 | 4 | 485 | ||
NR_024581.1 | 3 | 334 | ||||
NR_037164.1 | 4 | 485 | ||||
GenBank mRNA | AF403031.1 | - | 4 | 467 | ||
AK023196.1 | - | 4 | 468 | |||
AK091935.1 | - | 3 | 334 | |||
AK292071.1 | - | 4 | 471 | |||
BC012056.1 | - | 4 | 462 | 4552643 | ||
BG436652.1 | - | 3 | 311 | |||
BP249999.1 | - | 4 | 485 | |||
CR457302.1 | - | 4 | 441 |
