siRNA Details
Gene Symbol: ATAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190724.2 | NP_001177653.1 | 9 | 1253 | ||
NM_001318762.1 | NP_001305691.1 | 9 | 1015 | |||
NM_001318763.1 | NP_001305692.1 | 10 | 1084 | |||
GenBank mRNA | AK023220.1 | - | 10 | 1050 | ||
AK297661.1 | - | 9 | 1136 | |||
BC006105.1 | - | 9 | 1026 | 2820493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
