siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102366.2 | NP_001095836.1 | 16 | 2621 | ||
NM_001102367.2 | NP_001095837.1 | 15 | 2387 | |||
NM_001102368.2 | NP_001095838.1 | 14 | 2220 | |||
NM_001102369.2 | NP_001095839.1 | 15 | 2220 | |||
NM_001286470.1 | NP_001273399.1 | 17 | 2915 | |||
NM_001286471.1 | NP_001273400.1 | 14 | 2456 | |||
NM_001286472.1 | NP_001273401.1 | 15 | 2271 | |||
NM_024952.7 | NP_079228.4 | 14 | 2450 | |||
GenBank mRNA | AB210031.1 | - | 15 | 2121 | ||
AK022564.1 | - | 14 | 2021 | |||
AK023200.1 | - | 15 | 2018 | |||
AK024603.1 | - | 8 | 1248 | |||
AK091394.1 | - | 15 | 1810 | |||
AK097262.1 | - | 1 | 2184 | |||
AK097294.1 | - | 14 | 2170 | |||
AK098752.1 | - | 14 | 2171 | |||
AK123532.1 | - | 17 | 2558 | |||
AK222531.1 | - | 8 | 1246 | |||
AY358516.1 | - | 14 | 2456 | |||
BC010614.2 | - | 15 | 2336 | 4157432 | ||
BC065558.1 | - | 16 | 2244 | 6138321 | ||
BX161393.1 | - | 14 | 2149 | |||
BX247989.1 | - | 15 | 2199 | |||
BX248294.1 | - | 13 | 1939 | |||
BX537970.1 | - | 15 | 2167 | |||
BX647492.1 | - | 1 | 3635 | |||
DR003467.1 | - | 1 | 97 |
