siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102366.2 | NP_001095836.1 | 12 | 1969 | ||
NM_001102367.2 | NP_001095837.1 | 11 | 1735 | |||
NM_001102368.2 | NP_001095838.1 | 10 | 1568 | |||
NM_001102369.2 | NP_001095839.1 | 12 | 1688 | |||
NM_001286470.1 | NP_001273399.1 | 13 | 2263 | |||
NM_001286471.1 | NP_001273400.1 | 10 | 1804 | |||
NM_001286472.1 | NP_001273401.1 | 12 | 1739 | |||
NM_024952.7 | NP_079228.4 | 10 | 1798 | |||
GenBank mRNA | AB210031.1 | - | 11 | 1469 | ||
AK022564.1 | - | 10 | 1369 | |||
AK023200.1 | - | 12 | 1486 | |||
AK024603.1 | - | 4 | 594 | |||
AK091394.1 | - | 11 | 1158 | |||
AK097294.1 | - | 10 | 1518 | |||
AK098752.1 | - | 10 | 1519 | |||
AK123532.1 | - | 13 | 1906 | |||
AK222531.1 | - | 4 | 594 | |||
AY358516.1 | - | 10 | 1804 | |||
BC010614.2 | - | 11 | 1684 | 4157432 | ||
BC065558.1 | - | 12 | 1592 | 6138321 | ||
BX161393.1 | - | 10 | 1497 | |||
BX247989.1 | - | 12 | 1667 | |||
BX248294.1 | - | 8 | 1170 | |||
BX248782.1 | - | 11 | 1518 | |||
BX537970.1 | - | 10 | 1515 | |||
CR457289.1 | - | 8 | 1216 |
