siRNA Details
Gene Symbol: UBE2G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003342.4 | NP_003333.1 | 3 | 520 | ||
GenBank mRNA | AK313059.1 | - | 3 | 367 | ||
BC002775.2 | - | 3 | 419 | 3619626 | ||
BC026288.1 | - | 3 | 328 | |||
BT007416.1 | - | 3 | 162 | |||
D78514.1 | - | 3 | 180 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025985, AK013902, AK167472, BC065083, BC096474, and others.
Gene Ontology Categories:
