siRNA Details
Gene Symbol: UBE2N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003348.3 | NP_003339.1 | 2 | 522 | ||
GenBank mRNA | AK098233.1 | - | 1 | 125 | ||
BC000396.2 | - | 2 | 248 | 2822013 | ||
BC003365.1 | - | 2 | 183 | 2900313 | ||
BC108704.1 | - | 2 | 217 | 6503770 | ||
BI459891.1 | - | 2 | 539 | |||
BT006873.1 | - | 2 | 156 | |||
D83004.1 | - | 2 | 219 | |||
FJ224354.1 | - | 2 | 219 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_080560, BC067069, AK005788, BC052203, BC138211, and others.
Gene Ontology Categories:
