siRNA Details
Gene Symbol: UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032288.2 | NP_001027459.1 | 4 | 638 | ||
NM_001257393.1 | NP_001244322.1 | 5 | 813 | |||
NM_001257394.1 | NP_001244323.1 | 3 | 872 | |||
NM_001257395.1 | NP_001244324.1 | 4 | 629 | |||
NM_001257396.1 | NP_001244325.1 | 3 | 512 | |||
NM_001257397.1 | NP_001244326.1 | 5 | 721 | |||
NM_001257398.1 | NP_001244327.1 | 5 | 777 | |||
NM_001257399.1 | NP_001244328.1 | 5 | 756 | |||
NM_001282575.2 | NP_001269504.1 | 5 | 1073 | |||
NM_001282576.1 | NP_001269505.1 | 5 | 912 | |||
NM_001282577.2 | NP_001269506.1 | 5 | 690 | |||
NM_001282578.2 | NP_001269507.1 | 4 | 827 | |||
NM_001282579.1 | NP_001269508.1 | 5 | 738 | |||
NM_021988.5 | NP_068823.2 | 4 | 1021 | |||
NM_022442.5 | NP_071887.1 | 4 | 664 | |||
NM_199144.2 | NP_954595.1 | 5 | 944 | |||
NR_047554.1 | 3 | 489 | ||||
GenBank mRNA | AL110132.1 | - | 2 | 2653 | ||
BC000468.2 | - | 4 | 578 | 2961017 | ||
BC005215.1 | - | 5 | 1071 | |||
BC008944.2 | - | 3 | 555 | |||
BU934624.1 | - | 4 | 609 | |||
BX647511.1 | - | 3 | 457 | |||
BX648147.1 | - | 3 | 463 | |||
DA696213.1 | - | 3 | 479 | |||
U39360.1 | - | 5 | 706 | |||
U97279.1 | - | 1 | 905 |
Gene Symbol: TMEM189-UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199203.2 | NP_954673.1 | 8 | 1398 | ||
GenBank mRNA | BQ223768.1 | - | 1 | 143 | ||
BU156219.1 | - | 4 | 573 | |||
BU619725.1 | - | 2 | 371 | |||
CF145300.1 | - | 2 | 300 | |||
U39361.1 | - | 5 | 859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
