siRNA Details
Gene Symbol: MIR600HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026677.1 | 1 | 4835 | |||
GenBank mRNA | AF251293.1 | - | 1 | 524 | ||
AK025814.1 | - | 1 | 1997 | |||
AK123844.1 | - | 1 | 2565 | |||
BC015678.2 | - | 1 | 2825 | |||
BC034918.1 | - | 1 | 787 | |||
BX648949.1 | - | 1 | 4482 | |||
CR749230.1 | - | 1 | 4840 |
