siRNA Details
Gene Symbol: VAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134398.1 | NP_001127870.1 | 3, 4 | 427 | ||
NM_003371.3 | NP_003362.2 | 3, 4 | 427 | |||
XM_005272213.1 | XP_005272270.1 | 3, 4 | 477 | |||
XM_017015108.1 | XP_016870597.1 | 3, 4 | 477 | |||
XM_017015109.1 | XP_016870598.1 | 3, 4 | 477 | |||
XM_017015110.1 | XP_016870599.1 | 3, 4 | 477 | |||
XM_017015111.1 | XP_016870600.1 | 3, 4 | 477 | |||
XM_017015112.1 | XP_016870601.1 | 3, 4 | 477 | |||
XM_017015113.1 | XP_016870602.1 | 3, 4 | 477 | |||
GenBank mRNA | BC132965.1 | - | 3, 4 | 427 | 40146987 | |
BC132967.1 | - | 3, 4 | 427 | 40146989 | ||
BX640754.1 | - | 3, 4 | 267 | |||
S76992.1 | - | 3, 4 | 386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
