siRNA Details
Gene Symbol: CDCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031942.4 | NP_114148.3 | 7 | 1077 | ||
NM_145810.2 | NP_665809.1 | 6 | 840 | |||
XM_011511957.2 | XP_011510259.1 | 7 | 1618 | |||
GenBank mRNA | AK027628.1 | - | 7 | 1049 | ||
AK027642.1 | - | 6 | 821 | |||
AK075134.1 | - | 5 | 1703 | |||
AK223524.1 | - | 6 | 845 | |||
AK297097.1 | - | 5 | 675 | |||
AK297245.1 | - | 7 | 937 | |||
AK300949.1 | - | 7 | 906 | |||
AL833728.1 | - | 2 | 369 | |||
AL834186.1 | - | 5 | 1682 | |||
AY029179.1 | - | 6 | 853 | |||
BC015124.1 | - | 2 | 256 | |||
BC027966.1 | - | 6 | 806 | 5227112 |
