siRNA Details
Gene Symbol: ARMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032131.5 | NP_115507.4 | 3 | 453 | ||
XM_005267154.3 | XP_005267211.1 | 3 | 734 | |||
XM_005267155.2 | XP_005267212.1 | 3 | 469 | |||
XM_005267157.3 | XP_005267214.1 | 3 | 459 | |||
XM_006715573.2 | XP_006715636.1 | 4 | 559 | |||
XM_006715574.2 | XP_006715637.1 | 1 | 61 | |||
XM_011536166.1 | XP_011534468.1 | 4 | 544 | |||
XM_011536167.1 | XP_011534469.1 | 3 | 447 | |||
XM_011536168.2 | XP_011534470.1 | 3 | 447 | |||
XM_011536170.2 | XP_011534472.1 | 3 | 447 | |||
XM_011536176.2 | XP_011534478.1 | 3 | 447 | |||
XM_017011350.1 | XP_016866839.1 | 3 | 459 | |||
XM_017011351.1 | XP_016866840.1 | 3 | 447 | |||
XM_017011352.1 | XP_016866841.1 | 3 | 459 | |||
GenBank mRNA | AK095234.1 | - | 2 | 296 | ||
AK292499.1 | - | 3 | 447 | |||
BC030603.2 | - | 3 | 447 | 4824783 |
