siRNA Details
Gene Symbol: ZNF124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297568.1 | NP_001284497.1 | 4 | 471 | ||
XM_006711813.3 | XP_006711876.1 | 4 | 524 | |||
XM_011544272.2 | XP_011542574.1 | 4 | 322 | |||
GenBank mRNA | BC096351.2 | - | 4 | 421 | 40009547 | |
BC099661.3 | - | 4 | 421 | 40009541 | ||
S54641.1 | - | 1 | 676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
