siRNA Details
Gene Symbol: FAXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032511.2 | NP_115900.1 | 4 | 1009 | ||
XM_006715582.2 | XP_006715645.1 | 2 | 209 | |||
XM_011536188.2 | XP_011534490.1 | 4 | 526 | |||
GenBank mRNA | AK057793.1 | - | 3 | 427 | ||
AK096480.1 | - | 4 | 772 | |||
BC004869.2 | - | 4 | 1009 | 3834272 | ||
BC011583.1 | - | 4 | 1009 | 4577456 | ||
BQ883579.1 | - | 3 | 451 |
