siRNA Details
Gene Symbol: ZNF207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032293.2 | NP_001027464.1 | 3 | 406 | ||
NM_001098507.1 | NP_001091977.1 | 3 | 406 | |||
NM_003457.3 | NP_003448.1 | 3 | 406 | |||
XM_011525185.2 | XP_011523487.1 | 3 | 538 | |||
XM_017025019.1 | XP_016880508.1 | 3 | 427 | |||
XM_017025020.1 | XP_016880509.1 | 3 | 427 | |||
GenBank mRNA | AB209215.1 | - | 3 | 364 | ||
AF046001.1 | - | 3 | 460 | |||
AK226172.1 | - | 3 | 379 | |||
AK291801.1 | - | 3 | 369 | |||
AK298950.1 | - | 3 | 380 | |||
AK309979.1 | - | 4 | 577 | |||
BC002372.2 | - | 3 | 406 | 2822814 |
