siRNA Details
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 15 | 1786 | ||
NM_032830.2 | NP_116219.2 | 15 | 1840 | |||
XM_005256205.3 | XP_005256262.2 | 11 | 1371 | |||
GenBank mRNA | AB075868.2 | - | 15 | 1778 | ||
AK027419.1 | - | 12 | 1395 | |||
AK027445.1 | - | 10 | 1118 | |||
AK027584.1 | - | 4 | 417 | |||
AK027634.1 | - | 15 | 1792 | |||
AK027675.1 | - | 15 | 1789 | |||
AK074795.1 | - | 15 | 1806 | |||
BC000167.2 | - | 11 | 1285 | |||
BC009348.2 | - | 15 | 1772 | 4131625 | ||
BX647265.1 | - | 15 | 1754 |
