siRNA Details
Gene Symbol: CSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033225.5 | NP_150094.5 | 47 | 7461 | ||
XM_011534752.2 | XP_011533054.1 | 47 | 7690 | |||
XM_011534753.2 | XP_011533055.1 | 30 | 4568 | |||
XM_011534754.1 | XP_011533056.1 | 23 | 3304 | |||
XM_017013731.1 | XP_016869220.1 | 47 | 7690 | |||
GenBank mRNA | AF333704.2 | - | 48 | 7467 | ||
AY017307.1 | - | 48 | 7362 |
