siRNA Details
Gene Symbol: LOC494141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026541.1 | 2 | 1039 | |||
NR_026563.1 | 1 | 1184 | ||||
NR_026564.1 | 2 | 1039 | ||||
GenBank mRNA | AK131059.1 | - | 2 | 1017 | ||
BC026079.1 | - | 1 | 1191 | |||
BC026082.1 | - | 2 | 1036 | |||
BC034967.1 | - | 1 | 1191 | |||
BC044592.1 | - | 1 | 1184 | |||
BC056262.1 | - | 2 | 1041 |
Gene Symbol: SLC25A51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033412.3 | NP_219480.1 | 3 | 1239 | ||
GenBank mRNA | BC008500.1 | - | 3 | 1000 | 4286914 | |
BG953457.1 | - | 1 | 200 |
Gene Symbol: SLC25A51P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026540.1 | 1 | 1322 | |||
GenBank mRNA | BC024198.1 | - | 1 | 1323 |
Gene Symbol: SLC25A52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034172.2 | NP_001029344.3 | 1 | 1008 | ||
GenBank mRNA | AK302337.1 | - | 1 | 1008 | ||
BC103999.1 | - | 1 | 864 | 40023323 | ||
BC104000.1 | - | 1 | 864 | 40023324 | ||
BC104001.1 | - | 1 | 864 | 40023325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
