siRNA Details
Gene Symbol: BMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003940.1 | NP_001003940.1 | 4 | 628 | ||
NM_033503.3 | NP_277038.1 | 3 | 479 | |||
GenBank mRNA | AK024472.1 | - | 3 | 489 | ||
AY029254.1 | - | 2 | 393 | |||
BC060783.1 | - | 4 | 626 | |||
BC069328.1 | - | 3 | 517 | 7262161 | ||
BC069505.1 | - | 3 | 463 | 7262173 | ||
BC070043.1 | - | 4 | 626 | 30344794 | ||
BC104983.1 | - | 3 | 414 | 8143986 | ||
BC104985.1 | - | 3 | 414 | 8143988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
