siRNA Details
Gene Symbol: ZFP62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172638.1 | NP_001166109.1 | 2 | 2726 | ||
NM_152283.4 | NP_689496.4 | 3 | 2799 | |||
XM_017009710.1 | XP_016865199.1 | 3 | 2829 | |||
XM_017009711.1 | XP_016865200.1 | 3 | 2877 | |||
XM_017009712.1 | XP_016865201.1 | 4 | 2902 | |||
XM_017009713.1 | XP_016865202.1 | 4 | 2950 | |||
XM_017009714.1 | XP_016865203.1 | 3 | 2883 | |||
XM_017009715.1 | XP_016865204.1 | 2 | 2836 | |||
XM_017009717.1 | XP_016865206.1 | 2 | 2799 | |||
GenBank mRNA | AK002206.1 | - | 1 | 1282 | ||
AK091550.1 | - | 2 | 1556 | |||
AK295714.1 | - | 3 | 2789 | |||
AK303450.1 | - | 2 | 2717 | |||
AL832408.1 | - | 1 | 2886 | |||
BC140761.1 | - | 1 | 2105 | 9021629 | ||
BC171780.1 | - | 1 | 2105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
