siRNA Details
Gene Symbol: DIDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193369.1 | NP_001180298.1 | 16 | 7075 | ||
NM_033081.2 | NP_149072.2 | 16 | 7119 | |||
XM_011528504.1 | XP_011526806.1 | 17 | 7160 | |||
XM_011528505.1 | XP_011526807.1 | 17 | 7227 | |||
XM_011528506.1 | XP_011526808.1 | 17 | 14011 | |||
XM_011528507.1 | XP_011526809.1 | 17 | 7095 | |||
XM_011528508.1 | XP_011526810.1 | 17 | 7236 | |||
GenBank mRNA | AL133063.1 | - | 1 | 1899 | ||
AY481572.1 | - | 16 | 7027 | |||
BC012757.2 | - | 1 | 2811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
