siRNA Details
Gene Symbol: UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032288.2 | NP_001027459.1 | 4 | 1456 | ||
NM_001257393.1 | NP_001244322.1 | 5 | 1631 | |||
NM_001257394.1 | NP_001244323.1 | 3 | 1690 | |||
NM_001257395.1 | NP_001244324.1 | 4 | 1447 | |||
NM_001257396.1 | NP_001244325.1 | 3 | 1330 | |||
NM_001257397.1 | NP_001244326.1 | 5 | 1539 | |||
NM_001257398.1 | NP_001244327.1 | 5 | 1595 | |||
NM_001257399.1 | NP_001244328.1 | 5 | 1574 | |||
NM_001282575.2 | NP_001269504.1 | 5 | 1891 | |||
NM_001282576.1 | NP_001269505.1 | 5 | 1730 | |||
NM_001282577.2 | NP_001269506.1 | 5 | 1508 | |||
NM_001282578.2 | NP_001269507.1 | 4 | 1645 | |||
NM_001282579.1 | NP_001269508.1 | 5 | 1556 | |||
NM_021988.5 | NP_068823.2 | 4 | 1839 | |||
NM_022442.5 | NP_071887.1 | 4 | 1482 | |||
NM_199144.2 | NP_954595.1 | 5 | 1762 | |||
NR_047554.1 | 3 | 1307 | ||||
GenBank mRNA | AF049671.1 | - | 1 | 205 | ||
AL110132.1 | - | 2 | 3471 | |||
BC000468.2 | - | 4 | 1396 | 2961017 | ||
BC008944.2 | - | 3 | 1373 | |||
BX647511.1 | - | 3 | 1275 | |||
BX648147.1 | - | 3 | 1282 | |||
U39360.1 | - | 5 | 1524 | |||
U97279.1 | - | 1 | 1723 |
Gene Symbol: TMEM189-UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199203.2 | NP_954673.1 | 8 | 2216 | ||
GenBank mRNA | BM556348.1 | - | 1 | 276 | ||
BU860198.1 | - | 1 | 578 | |||
U39361.1 | - | 5 | 1677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
