siRNA Details
Gene Symbol: ROPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317774.1 | NP_001304703.1 | 3 | 474 | ||
NM_001317775.1 | NP_001304704.1 | 2 | 346 | |||
NM_017578.3 | NP_060048.2 | 4 | 554 | |||
XM_011512933.2 | XP_011511235.1 | 4 | 518 | |||
XM_011512934.2 | XP_011511236.1 | 4 | 629 | |||
GenBank mRNA | AF303889.2 | - | 4 | 554 | ||
BC132744.1 | - | 2 | 310 | 40146766 | ||
BC132746.1 | - | 2 | 311 | 40146768 | ||
BG722939.1 | - | 2 | 306 | |||
BG724071.1 | - | 3 | 442 |
Gene Symbol: ROPN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012337.2 | NP_001012337.1 | 3 | 482 | ||
NM_001308313.1 | NP_001295242.1 | 4 | 528 | |||
XM_005247138.3 | XP_005247195.1 | 2 | 337 | |||
XM_006713513.3 | XP_006713576.1 | 5 | 624 | |||
GenBank mRNA | AF231410.1 | - | 3 | 441 | ||
AK302014.1 | - | 3 | 387 | |||
BC015413.1 | - | 1 | 36 | 4394326 | ||
BC141849.1 | - | 4 | 475 | 8860347 | ||
DC397187.1 | - | 4 | 528 | |||
HY006578.1 | - | 3 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
