siRNA Details
Gene Symbol: DNAJB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017626.4 | NP_060096.3 | 8, 9 | 1408 | ||
GenBank mRNA | AK000034.1 | - | 8, 9 | 1184 | ||
AK225051.1 | - | 8, 9 | 1184 | |||
BC064920.1 | - | 8, 9 | 1174 | 6092812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
