siRNA Details
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017677.3 | NP_060147.2 | 4 | 491 | ||
GenBank mRNA | AK000133.1 | - | 2 | 150 | ||
AK225231.1 | - | 2 | 150 | |||
BC012399.1 | - | 4 | 469 | 3864128 |
