siRNA Details
Gene Symbol: NHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017838.3 | NP_060308.1 | 2, 3 | 380 | ||
GenBank mRNA | AF161404.1 | - | 2, 3 | 290 | ||
AF401219.1 | - | 2, 3 | 238 | |||
AJ293309.1 | - | 2, 3 | 304 | |||
AK000486.1 | - | 2, 3 | 286 | |||
BC000009.2 | - | 2, 3 | 288 | 3505294 | ||
BC006387.2 | - | 2, 3 | 314 | 4109843 | ||
BE797477.1 | - | 2, 3 | 400 | |||
BG827491.1 | - | 2, 3 | 388 | |||
CR457238.1 | - | 2, 3 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
