siRNA Details
Gene Symbol: EPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017957.2 | NP_060427.2 | 5, 6 | 1319 | ||
GenBank mRNA | AF324241.1 | - | 4, 5 | 884 | ||
AK000785.1 | - | 5, 6 | 1268 | |||
AK291658.1 | - | 5, 6 | 1268 | |||
AK292977.1 | - | 5, 6 | 1268 | |||
AK296346.1 | - | 5, 6 | 1105 | |||
AK300642.1 | - | 6, 7 | 767 | |||
BC110602.2 | - | 4, 5 | 851 |
