siRNA Details
Gene Symbol: RBM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018047.2 | NP_060517.1 | 7 | 766 | ||
GenBank mRNA | AK001152.1 | - | 7 | 721 | ||
AK297019.1 | - | 6 | 535 | |||
AL136933.1 | - | 7 | 695 | |||
BC003402.1 | - | 7 | 699 | 3449258 |
