siRNA Details
Gene Symbol: VRTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018228.2 | NP_060698.2 | 2 | 1386 | ||
XM_011536911.1 | XP_011535213.1 | 3 | 1433 | |||
GenBank mRNA | AK001673.1 | - | 2 | 1340 | ||
AK023213.1 | - | 2 | 1353 | |||
BC053325.1 | - | 2 | 1369 | 6577658 |
