siRNA Details
Gene Symbol: ZNF839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267827.1 | NP_001254756.1 | 8 | 1986 | ||
NM_001267828.1 | NP_001254757.1 | 8 | 1917 | |||
NM_018335.4 | NP_060805.3 | 8 | 1999 | |||
XM_006720203.3 | XP_006720266.2 | 8 | 1883 | |||
XM_011536945.1 | XP_011535247.1 | 9 | 2101 | |||
XM_011536946.2 | XP_011535248.1 | 8 | 1951 | |||
XM_011536948.2 | XP_011535250.1 | 8 | 1727 | |||
XM_017021447.1 | XP_016876936.1 | 7 | 1849 | |||
XM_017021449.1 | XP_016876938.1 | 7 | 1265 | |||
GenBank mRNA | AF155112.1 | - | 6 | 628 | ||
AK001994.1 | - | 4 | 415 | |||
AK093342.1 | - | 8 | 1986 | |||
AK125678.1 | - | 4 | 1135 | |||
AK127821.1 | - | 4 | 1233 | |||
AK223312.1 | - | 4 | 399 | |||
AK289632.1 | - | 8 | 1917 | |||
BC073137.1 | - | 6 | 782 |
