siRNA Details
Gene Symbol: CDK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098533.2 | NP_001092003.2 | 2 | 154 | ||
NM_001160367.1 | NP_001153839.1 | 2 | 154 | |||
NM_052987.3 | NP_443713.2 | 2 | 154 | |||
NM_052988.4 | NP_443714.3 | 2 | 192 | |||
XM_006721308.2 | XP_006721371.1 | 2 | 248 | |||
XM_006721310.2 | XP_006721373.1 | 2 | 248 | |||
XM_011523405.2 | XP_011521707.1 | 2 | 248 | |||
XM_011523406.2 | XP_011521708.1 | 2 | 248 | |||
XM_011523407.2 | XP_011521709.1 | 2 | 248 | |||
XM_011523408.2 | XP_011521710.1 | 2 | 248 | |||
XM_011523410.2 | XP_011521712.1 | 2 | 166 | |||
XM_011523411.2 | XP_011521713.1 | 3 | 404 | |||
XM_011523412.1 | XP_011521714.1 | 3 | 374 | |||
XM_011523413.2 | XP_011521715.1 | 2 | 806 | |||
XM_011523414.2 | XP_011521716.1 | 2 | 592 | |||
XM_011523417.2 | XP_011521719.1 | 2 | 248 | |||
XM_017023807.1 | XP_016879296.1 | 2 | 248 | |||
XM_017023808.1 | XP_016879297.1 | 3 | 403 | |||
XM_017023810.1 | XP_016879299.1 | 2 | 591 | |||
GenBank mRNA | AK292351.1 | - | 2 | 170 | ||
AK298386.1 | - | 2 | 192 | |||
BC017342.2 | - | 2 | 134 | 4865641 | ||
BC093028.1 | - | 2 | 171 | |||
CX867290.1 | - | 2 | 157 | |||
DC362478.1 | - | 2 | 192 | |||
L33264.1 | - | 2 | 153 | |||
X78342.1 | - | 2 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
