siRNA Details
Gene Symbol: DOK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018431.4 | NP_060901.2 | 8 | 1826 | ||
NM_177959.2 | NP_808874.1 | 8 | 1472 | |||
XM_011528904.1 | XP_011527206.1 | 8 | 1455 | |||
GenBank mRNA | AF132732.1 | - | 8 | 1548 | ||
AL050069.1 | - | 7 | 1229 | |||
BI916998.1 | - | 172 | ||||
CN427887.1 | - | 1 | 155 |
