siRNA Details
Gene Symbol: ZKSCAN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288590.1 | NP_001275519.1 | 6 | 2799 | ||
NM_018651.3 | NP_061121.2 | 6 | 2836 | |||
GenBank mRNA | AF154846.1 | - | 5 | 2452 | ||
AK291285.1 | - | 6 | 2781 | |||
AY280798.1 | - | 6 | 2790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
