siRNA Details
Gene Symbol: BRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018963.4 | NP_061836.2 | 17 | 2108 | ||
NM_033656.3 | NP_387505.1 | 17 | 2108 | |||
XM_011529611.1 | XP_011527913.1 | 17 | 2141 | |||
XM_011529612.2 | XP_011527914.1 | 10 | 1004 | |||
XM_017028373.1 | XP_016883862.1 | 18 | 1913 | |||
XM_017028375.1 | XP_016883864.1 | 17 | 2141 | |||
GenBank mRNA | AB080586.1 | - | 17 | 1912 | ||
AB080587.1 | - | 17 | 1912 | |||
AB080588.1 | - | 9 | 947 | |||
AB080589.1 | - | 9 | 947 | |||
AB080591.1 | - | 9 | 947 | |||
AB080592.1 | - | 9 | 852 | |||
AB080593.1 | - | 9 | 826 | |||
AJ238214.2 | - | 17 | 1912 | |||
AJ292465.1 | - | 17 | 1892 | |||
AJ292466.1 | - | 17 | 1892 | |||
AK125906.1 | - | 2 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
