siRNA Details
Gene Symbol: SNTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287813.2 | NP_001274742.1 | 20 | 2465 | ||
NM_001287814.2 | NP_001274743.1 | 17 | 2506 | |||
NM_001321773.1 | NP_001308702.1 | 18 | 2617 | |||
NM_018967.4 | NP_061840.1 | 19 | 2692 | |||
NR_135794.1 | 19 | 2672 | ||||
NR_135795.1 | 19 | 2687 | ||||
NR_135796.1 | 19 | 2718 | ||||
NR_135797.1 | 21 | 2862 | ||||
XM_011517548.2 | XP_011515850.1 | 18 | 2802 | |||
XM_017013579.1 | XP_016869068.1 | 21 | 5621 | |||
XM_017013580.1 | XP_016869069.1 | 19 | 2215 | |||
GenBank mRNA | AL161971.1 | - | 19 | 2587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
