siRNA Details
Gene Symbol: HIST1H4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003546.2 | NP_003537.1 | 1 | 154 | ||
GenBank mRNA | AB451459.1 | - | 1 | 153 | ||
BC069392.1 | - | 1 | 154 | 7262681 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HIST1H4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003546.2 | NP_003537.1 | 1 | 154 | ||
GenBank mRNA | AB451459.1 | - | 1 | 153 | ||
BC069392.1 | - | 1 | 154 | 7262681 |