siRNA Details
Gene Symbol: NPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003995.3 | NP_003986.2 | 2, 3 | 872 | ||
XM_005251478.3 | XP_005251535.1 | 2, 3 | 1659 | |||
GenBank mRNA | AJ005282.1 | - | 2, 3 | 872 | ||
L13436.1 | - | 2, 3 | 1522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
