siRNA Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004720.2 | NP_001004720.1 | 3 | 810 | ||
NM_003581.4 | NP_003572.2 | 4 | 920 | |||
XM_006712797.3 | XP_006712860.1 | 4 | 1576 | |||
XM_011511991.2 | XP_011510293.1 | 4 | 1014 | |||
XM_017005103.1 | XP_016860592.1 | 5 | 1178 | |||
XM_017005104.1 | XP_016860593.1 | 4 | 1190 | |||
XM_017005105.1 | XP_016860594.1 | 4 | 927 | |||
GenBank mRNA | AF043119.1 | - | 2 | 672 | ||
AF047487.1 | - | 3 | 776 | |||
BC000103.2 | - | 3 | 694 | 3505427 | ||
BC007195.1 | - | 3 | 701 | 3608149 | ||
KU178418.1 | - | 2 | 644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
