siRNA Details
Gene Symbol: SMIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042493.2 | NP_001035958.1 | 4 | 239 | ||
NM_020425.5 | NP_065158.3 | 3 | 218 | |||
GenBank mRNA | AF311339.1 | - | 3 | 260 | ||
AK311963.1 | - | 4 | 210 | |||
AL050201.1 | - | 2 | 185 | |||
BC070260.1 | - | 3 | 200 | 6452586 | ||
BG534658.1 | - | 3 | 219 | |||
DA293899.1 | - | 4 | 210 |
